| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10500361 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10500360 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0300019618.mm.1 |
NR_040409.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019618.mm.1 |
NR_040409.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019618.mm.1 |
TC0300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019612.mm.1 |
NR_040409.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019612.mm.1 |
NR_040409.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019612.mm.1 |
TC0300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019619.mm.1 |
NR_040409.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019619.mm.1 |
NR_040409.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019619.mm.1 |
TC0300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019616.mm.1 |
NR_040409.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019616.mm.1 |
NR_040409.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019616.mm.1 |
TC0300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019614.mm.1 |
NR_040409.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019614.mm.1 |
NR_040409.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019614.mm.1 |
TC0300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019613.mm.1 |
NR_040409.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019613.mm.1 |
NR_040409.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019613.mm.1 |
TC0300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019617.mm.1 |
NR_040409.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019617.mm.1 |
NR_040409.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019617.mm.1 |
TC0300002520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019620.mm.1 |
NR_040409.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019620.mm.1 |
NR_040409.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019620.mm.1 |
TC0300002520.mm.1 |
MTA TC |