Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04336676_m1
          See other PRLR GE Assays ›
          Gene Symbol:
          Prlr
          Gene Name:
          prolactin receptor
          Gene Aliases:
          AI987712, Pr-1, Pr-3, Prlr-rs1
          Chromosome Location:
          Chr.15: 10177238 - 10349190 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04336676_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prlr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253781.1 NP_001240710.1 4 - 5 491 84
          NM_001253782.2 NP_001240711.1 3 - 4 300 84
          NM_011169.5 NP_035299.4 4 - 5 934 84
          GenBank mRNA AB643813.1 - 2 - 3 260 84
          AK050317.1 - 4 - 5 526 84
          AK052679.1 - 4 - 5 389 84
          AK054381.1 - 4 - 5 754 84
          AK054551.1 - 4 - 5 935 84
          AK081676.1 - 4 - 5 392 84
          AK143335.1 - 4 - 5 621 84
          BC005555.1 - 4 - 5 517 3586308 84
          BC006652.1 - 4 - 5 408 3500076 84
          BP761592.1 - 4 - 5 517 84
          D10214.1 - 2 - 3 209 84
          L13593.1 - 2 - 3 244 84
          L14811.1 - 2 - 3 186 84
          M22957.1 - 4 - 5 412 84
          M22958.1 - 2 - 3 246 84
          M22959.1 - 4 - 5 412 84
          X73372.1 - 2 - 3 186 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002327.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prlr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of transmembrane receptor protein tyrosine kinase activity
          JAK-STAT cascade
          lactation
          regulation of cell adhesion
          regulation of epithelial cell differentiation
          prolactin signaling pathway
          mammary gland epithelial cell differentiation
          prostate gland growth
          mammary gland alveolus development
          mammary gland epithelium development
          cytokine receptor activity
          prolactin receptor activity
          protein binding
          peptide hormone binding
          protein kinase binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500000699.mm.1 NM_001253781.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000699.mm.1 NM_001253781.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000699.mm.1 NM_001253782.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000699.mm.1 NM_001253782.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000699.mm.1 NM_011169.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000699.mm.1 NM_011169.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000699.mm.1 TC1500002327.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 380085 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline