Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271498.1 | NP_001258427.1 | 4 - 5 | 716 | 63 | |
GenBank mRNA | AK040940.1 | - | 4 - 5 | 568 | 63 | |
AK080000.1 | - | 4 - 5 | 568 | 63 | ||
AY221619.1 | - | 4 - 5 | 455 | 63 | ||
BC145348.1 | - | 4 - 5 | 356 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |