Search
Search

Pi4kb| Gene Symbol: Pi4kb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293715.1 | NP_001280644.1 | 9 - 10 | 2391 | 89 | |
| NM_001293716.1 | NP_001280645.1 | 7 - 8 | 1409 | 89 | ||
| NM_175356.3 | NP_780565.2 | 8 - 9 | 2346 | 89 | ||
| GenBank mRNA | AK028974.1 | - | 8 - 9 | 2331 | 89 | |
| AK030704.1 | - | 8 - 9 | 2106 | 89 | ||
| AK048405.1 | - | 7 - 8 | 1202 | 89 | ||
| AK053614.1 | - | 8 - 9 | 2341 | 89 | ||
| AK157918.1 | - | 8 - 9 | 2138 | 89 | ||
| AK172074.1 | - | 8 - 9 | 2339 | 89 | ||
| BC079846.1 | - | 8 - 9 | 2346 | 5707855 | 89 | |
| BC113176.1 | - | 9 - 10 | 2246 | 40060773 | 89 | |
| BC113781.1 | - | 7 - 8 | 1309 | 40060770 | 89 | |
| BC138457.1 | - | 8 - 9 | 2249 | 8861477 | 89 | |
| BC138458.1 | - | 8 - 9 | 2249 | 8861478 | 89 | |
Set Membership: |
|