Search
Search

Vax2os| Gene Symbol: Vax2os | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_075095.1 | 2 - 3 | 1030 | 76 | |||
| GenBank mRNA | AY589788.1 | - | LIT3408 Vax2OS |
2 - 3 | 1027 | 76 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CD207 antigen | Cd207 | -24.6kb | Sense | Chr. 6 - 83671207 - 83677857 on NCBI Build GRCm38 | |
| Downstream | ventral anterior homeobox 2 | Vax2 | Dres93 | 7.2kb | Anti-sense | Chr. 6 - 83711264 - 83739229 on NCBI Build GRCm38 |
Set Membership: |
|