Search
Search

Rad51d| Gene Symbol: Rad51d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102717.1 | 7 - 8 | 878 | 88 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger and FYVE like domain containing protein | Rffl |
1700051E09Rik, 4930516L10Rik, BG080975, Carp2 |
-7.9kb | Sense | Chr. 11 - 82803819 - 82871239 on NCBI Build GRCm38 |
| Downstream | fibronectin type III domain containing 8 | Fndc8 | 4930466G16Rik | 10.3kb | Anti-sense | Chr. 11 - 82892145 - 82900737 on NCBI Build GRCm38 |
Set Membership: |
|