Search
Search

Plcd1| Gene Symbol: Plcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121571.1 | 9 - 10 | 1558 | 55 | ||
| GenBank mRNA | BC015249.1 | - | 9 - 10 | 1512 | 4241580 | 55 |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | villin-like | Vill | Villp | -2.2kb | Anti-sense | Chr. 9 - 119052729 - 119071525 on NCBI Build GRCm38 |
| Downstream | deleted in lung and esophageal cancer 1 | Dlec1 | D630005C06Rik | 27.2kb | Anti-sense | Chr. 9 - 119101535 - 119148246 on NCBI Build GRCm38 |
Set Membership: |
|