Search
Search
| Gene Symbol: Fdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301728.1 | NP_001288657.1 | 2 - 3 | 461 | 88 | |
| NM_007996.2 | NP_032022.1 | 2 - 3 | 461 | 88 | ||
| GenBank mRNA | AK004879.1 | - | 2 - 3 | 415 | 88 | |
| AK148115.1 | - | 2 - 3 | 416 | 88 | ||
| BC099518.1 | - | 2 - 3 | 315 | 88 | ||
| BC132173.1 | - | 2 - 3 | 386 | 40130450 | 88 | |
| BC132175.1 | - | 2 - 3 | 386 | 40130452 | 88 | |
| D43689.1 | - | 2 - 3 | 392 | 88 | ||
| D43690.1 | - | 2 - 3 | 387 | 88 | ||
| L29123.1 | - | 2 - 3 | 414 | 88 | ||
Set Membership: |
|