Search
Search
| Gene Symbol: Lep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008493.3 | NP_032519.1 | 1 - 2 | 32 | 71 | |
| GenBank mRNA | AK030984.1 | - | 1 - 2 | 32 | 71 | |
| AK142589.1 | - | 1 - 2 | 35 | 71 | ||
| HQ166716.1 | - | 1 - 2 | 32 | 71 | ||
| U18812.1 | - | 1 - 2 | 29 | 71 | ||
Set Membership: |
|