Search
Search

Haus5| Gene Symbol: Haus5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027999.1 | NP_082275.1 | 16 - 17 | 1590 | 106 | |
| GenBank mRNA | AK009476.1 | - | 16 - 17 | 1570 | 106 | |
| AK129228.1 | - | 12 - 13 | 2858 | 106 | ||
| AK153920.1 | - | 16 - 17 | 1573 | 106 | ||
| BC048724.1 | - | 2 - 3 | 215 | 106 | ||
| BC089002.1 | - | 16 - 17 | 1559 | 106 | ||
| BC118934.1 | - | 16 - 17 | 1565 | 40090953 | 106 | |
| BC118935.1 | - | 16 - 17 | 1565 | 40090954 | 106 | |
Set Membership: |
|