Search
Search

Setd8| Gene Symbol: Setd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030241.2 | NP_084517.2 | 1 - 2 | 90 | 127 | |
| GenBank mRNA | AK030904.1 | - | 1 - 2 | 83 | 128 | |
| BB619898.1 | - | 1 - 2 | 80 | 127 | ||
| BY041630.1 | - | 1 - 2 | 91 | 127 | ||
Set Membership: |
|