Search
Search
| Gene Symbol: Ghdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031871.1 | NP_114077.1 | 3 - 4 | 482 | 71 | |
| GenBank mRNA | AF316996.1 | - | 3 - 4 | 482 | 71 | |
| AK135322.1 | - | 3 - 4 | 484 | 71 | ||
| BC005639.1 | - | 3 - 4 | 463 | 3256858 | 71 | |
Set Membership: |
|