Search
Search
| Gene Symbol: Snx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080557.2 | NP_542124.1 | 10 - 11 | 952 | 91 | |
| GenBank mRNA | AK007706.1 | - | 10 - 11 | 994 | 91 | |
| AK030657.1 | - | 10 - 11 | 965 | 91 | ||
| AK031368.1 | - | 964 | 91 | |||
| AK135356.1 | - | 10 - 11 | 959 | 91 | ||
| BC006043.1 | - | 10 - 11 | 942 | 91 | ||
| BC016599.1 | - | 10 - 11 | 952 | 2644991 | 91 | |
Set Membership: |
|