Search
Search

Clrn1| Gene Symbol: Clrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153386.3 | NP_700435.1 | 1 - 2 | 420 | 95 | |
| GenBank mRNA | AF495720.1 | - | 1 - 2 | 418 | 95 | |
| AK041623.1 | - | 1 - 2 | 356 | 95 | ||
| AK042523.1 | - | 1 - 2 | 356 | 95 | ||
| BC132254.1 | - | 1 - 2 | 402 | 40130531 | 95 | |
| BC132256.1 | - | 1 - 2 | 402 | 40130533 | 95 | |
Set Membership: |
|