Search
Search

A830008E24Rik| Gene Symbol: A830008E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154552.1 | - | 1 - 2 | 515 | 78 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutamate-ammonia ligase (glutamine synthetase) | Glul |
GS, Glns |
-202.5kb | Anti-sense | Chr. 1 - 153899929 - 153909723 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 648 | Zfp648 |
ENSMUSG00000066797, Gm10178, Gm203 |
83.2kb | Anti-sense | Chr. 1 - 154201187 - 154212916 on NCBI Build GRCm38 |
Set Membership: |
|