Search
Search

OSBPL6| Gene Symbol: OSBPL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012132725.2 | XP_011988115.2 | 9 - 10 | 1144 | 77 | |
| XM_027964798.1 | XP_027820599.1 | 9 - 10 | 1546 | 77 | ||
| XM_027964799.1 | XP_027820600.1 | 12 - 13 | 1450 | 77 | ||
Set Membership: |
|