Search
Search

RPS6KC1| Gene Symbol: RPS6KC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013886.4 | XP_004013935.2 | 11 - 12 | 2879 | 85 | |
| XM_027956287.1 | XP_027812088.1 | 10 - 11 | 4282 | 85 | ||
| XM_027956288.1 | XP_027812089.1 | 11 - 12 | 2806 | 85 | ||
| XM_027956289.1 | XP_027812090.1 | 9 - 10 | 4166 | 85 | ||
| XM_027956290.1 | XP_027812091.1 | 4 - 5 | 2416 | 85 | ||
Set Membership: |
|