Search
Search

PSEN1| Gene Symbol: PSEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012181995.2 | XP_012037385.1 | 3 - 4 | 520 | 79 | |
| XM_012181996.3 | XP_012037386.1 | 3 - 4 | 570 | 79 | ||
| XM_012181998.3 | XP_012037388.1 | 2 - 3 | 393 | 79 | ||
Set Membership: |
|