Search
Search

PAPOLA| Gene Symbol: PAPOLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017986.4 | XP_004018035.1 | 17 - 18 | 1867 | 94 | |
| XM_012099023.3 | XP_011954413.1 | 17 - 18 | 1880 | 94 | ||
| XM_012099024.3 | XP_011954414.1 | 17 - 18 | 1879 | 94 | ||
| XM_012099025.3 | XP_011954415.1 | 17 - 18 | 1879 | 94 | ||
| XM_012099026.3 | XP_011954416.1 | 17 - 18 | 2972 | 94 | ||
Set Membership: |
|