Search
Search

ITIH4| Gene Symbol: ITIH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018391.4 | XP_004018440.2 | 5 - 6 | 764 | 60 | |
| XM_012099923.3 | XP_011955313.2 | 6 - 7 | 759 | 60 | ||
| XM_027958298.1 | XP_027814099.1 | 5 - 6 | 881 | 60 | ||
| XM_027958299.1 | XP_027814100.1 | 5 - 6 | 881 | 60 | ||
| XM_027958300.1 | XP_027814101.1 | 5 - 6 | 881 | 60 | ||
Set Membership: |
|