Search
Search

TADA2A| Gene Symbol: TADA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012185269.3 | XP_012040659.2 | 15 - 16 | 1416 | 61 | |
| XM_015098476.2 | XP_014953962.2 | 15 - 16 | 1318 | 61 | ||
| XM_015098480.2 | XP_014953966.2 | 15 - 16 | 1308 | 61 | ||
| XM_015098481.2 | XP_014953967.1 | 15 - 16 | 1332 | 61 | ||
Set Membership: |
|