Search
Search

ARMC8| Gene Symbol: ARMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092647.2 | XP_014948133.1 | 5 - 6 | 723 | 59 | |
| XM_015092648.2 | XP_014948134.1 | 5 - 6 | 728 | 59 | ||
| XM_027960071.1 | XP_027815872.1 | 5 - 6 | 729 | 59 | ||
| XM_027960080.1 | XP_027815881.1 | 4 - 5 | 652 | 59 | ||
| XM_027960081.1 | XP_027815882.1 | 5 - 6 | 734 | 59 | ||
Set Membership: |
|