Search
Search

EIF2B3| Gene Symbol: EIF2B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004001908.4 | XP_004001957.1 | 8 - 9 | 1041 | 70 | |
| XM_012124123.3 | XP_011979513.1 | 8 - 9 | 1060 | 70 | ||
Set Membership: |
|