Search
Search

PLEKHO1| Gene Symbol: PLEKHO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092222.2 | XP_014947708.2 | 3 - 4 | 364 | 64 | |
| XM_015092223.2 | XP_014947709.2 | 2 - 3 | 1791 | 64 | ||
| XM_015092224.2 | XP_014947710.1 | 3 - 4 | 575 | 64 | ||
Set Membership: |
|