Search
Search
| Gene Symbol: HPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977426.1 | XP_027833227.1 | 3 - 4 | 288 | 76 | |
| XM_027977427.1 | XP_027833228.1 | 3 - 4 | 288 | 76 | ||
| XM_027977428.1 | XP_027833229.1 | 3 - 4 | 306 | 76 | ||
Set Membership: |
|