Search
Search

ZRANB2
LOC101103178
LOC114118380| Gene Symbol: ZRANB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002081.4 | XP_004002130.3 | 2 - 3 | 157 | 116 | |
| XM_015092049.2 | XP_014947535.1 | 2 - 3 | 157 | 116 | ||
| Gene Symbol: LOC101103178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976325.1 | XP_027832126.1 | 1 | 192 | 116 | |
| XM_027976326.1 | XP_027832127.1 | 1 | 193 | 116 | ||
| Gene Symbol: LOC114118380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979158.1 | XP_027834959.1 | 1 | 138 | 116 | |
| XM_027979159.1 | XP_027834960.1 | 1 | 139 | 116 | ||
Set Membership: |
|