Search
Search
| Gene Symbol: HEYL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004001829.4 | XP_004001878.2 | 3 - 4 | 269 | 93 | |
| XM_012102566.3 | XP_011957956.2 | 3 - 4 | 444 | 93 | ||
| XM_012102640.3 | XP_011958030.2 | 3 - 4 | 196 | 93 | ||
Set Membership: |
|