Search
Search

SH3RF1| Gene Symbol: SH3RF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093285.2 | XP_014948771.2 | 4 - 5 | 945 | 80 | |
| XM_027964644.1 | XP_027820445.1 | 4 - 5 | 945 | 80 | ||
| XM_027964645.1 | XP_027820446.1 | 4 - 5 | 940 | 80 | ||
| XM_027964646.1 | XP_027820447.1 | 4 - 5 | 945 | 80 | ||