Search
Search

PRKAA1| Gene Symbol: PRKAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017019.3 | XP_004017068.2 | 5 - 6 | 601 | 84 | |
| XM_012097009.2 | XP_011952399.1 | 6 - 7 | 695 | 84 | ||
| XM_027980096.1 | XP_027835897.1 | 5 - 6 | 601 | 84 | ||
Set Membership: |
|