Search
Search

SLC24A5| Gene Symbol: SLC24A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252181.1 | NP_001239110.1 | 8 - 9 | 1183 | 72 | |
| GenBank mRNA | JF719290.1 | - | 8 - 9 | 1183 | 72 | |
Set Membership: |
|