Search
Search

WDFY4| Gene Symbol: WDFY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021557.4 | XP_004021606.2 | 7 - 8 | 1089 | 54 | |
| XM_012105527.3 | XP_011960917.2 | 7 - 8 | 1089 | 54 | ||
| XM_027962399.1 | XP_027818200.1 | 6 - 7 | 1026 | 54 | ||
Set Membership: |
|