Search
Search

THAP9| Gene Symbol: THAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096596.2 | XP_014952082.2 | 4 - 5 | 833 | 114 | |
| XM_015096597.2 | XP_014952083.2 | 4 - 5 | 898 | 114 | ||
Set Membership: |
|