Search
Search
| Gene Symbol: RIC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016178.4 | XP_004016227.2 | 3 - 4 | 421 | 91 | |
| XM_012095822.3 | XP_011951212.2 | 3 - 4 | 421 | 91 | ||
| XM_012095823.3 | XP_011951213.2 | 3 - 4 | 421 | 91 | ||
| XM_027979409.1 | XP_027835210.1 | 2 - 3 | 210 | 91 | ||