Search
Search
| Gene Symbol: OXTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009752.1 | NP_001009752.1 | 1 - 2 | 930 | 91 | |
| GenBank mRNA | AF344638.1 | - | 1 - 2 | 930 | 91 | |
| HM856597.1 | - | 1 - 2 | 930 | 91 | ||
| X87986.1 | - | 934 | 91 | |||
Set Membership: |
|