Search
Search

TIMELESS| Gene Symbol: TIMELESS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128153.1 | NP_001121625.1 | 25 - 26 | 3234 | 58 | |
| XM_027965745.1 | XP_027821546.1 | 26 - 27 | 3351 | 58 | ||
| XM_027965746.1 | XP_027821547.1 | 27 - 28 | 3616 | 58 | ||
| XM_027965747.1 | XP_027821548.1 | 27 - 28 | 3538 | 58 | ||
| GenBank mRNA | EF494244.1 | - | 25 - 26 | 3234 | 58 | |
Set Membership: |
|