Search
Search
| Gene Symbol: TTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009800.1 | NP_001009800.1 | 3 - 4 | 344 | 56 | |
| XM_027960684.1 | XP_027816485.1 | 3 - 4 | 370 | 56 | ||
| GenBank mRNA | X15576.1 | - | 344 | 56 | ||
Set Membership: |
|