Search
Search

PLAUR| Gene Symbol: PLAUR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163606.1 | NP_001157078.1 | 1 - 2 | 100 | 76 | |
| XM_012189516.3 | XP_012044906.2 | 1 - 2 | 76 | |||
| GenBank mRNA | FJ803200.1 | - | 1 - 2 | 100 | 76 | |
Set Membership: |
|