Search
Search

UBQLN1| Gene Symbol: UBQLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161886.1 | NP_001155358.1 | 6 - 7 | 1102 | 64 | |
| XM_012110487.3 | XP_011965877.1 | 6 - 7 | 1340 | 64 | ||
| GenBank mRNA | FJ958367.1 | - | 6 - 7 | 1102 | 64 | |
Set Membership: |
|