Search
Search

MADCAM1| Gene Symbol: MADCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038020.1 | NP_001033109.1 | 3 - 4 | 670 | 112 | |
| GenBank mRNA | DQ288287.1 | - | 3 - 4 | 670 | 112 | |
Set Membership: |
|