Search
Search
| Gene Symbol: TRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009407.1 | NP_001009407.1 | 2 - 3 | 1131 | 107 | |
| XM_027972753.1 | XP_027828554.1 | 1 - 2 | 793 | 107 | ||
| GenBank mRNA | X95285.1 | - | 1131 | 107 | ||
Set Membership: |
|