Search
Search
| Gene Symbol: FTO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104931.1 | NP_001098401.1 | 1 - 2 | 50 | 116 | |
| XM_027977360.1 | XP_027833161.1 | 1 - 2 | 147 | 116 | ||
| GenBank mRNA | EU072419.1 | - | 1 - 2 | 50 | 116 | |
Set Membership: |
|