Search
Search
| Gene Symbol: G6PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093780.1 | NP_001087249.1 | 3 - 4 | 223 | 73 | |
| XM_027962764.1 | XP_027818565.1 | 4 - 5 | 380 | 73 | ||
| XM_027962765.1 | XP_027818566.1 | 4 - 5 | 380 | 73 | ||
| XM_027962766.1 | XP_027818567.1 | 3 - 4 | 644 | 73 | ||
| XM_027962767.1 | XP_027818568.1 | 3 - 4 | 378 | 73 | ||
| XM_027962768.1 | XP_027818569.1 | 3 - 4 | 272 | 73 | ||
| XM_027962769.1 | XP_027818570.1 | 3 - 4 | 272 | 73 | ||
| GenBank mRNA | DQ377363.1 | - | 3 - 4 | 223 | 73 | |
| DQ377364.1 | - | 3 - 4 | 223 | 73 | ||
Set Membership: |
|