Search
Search

ACACA| Gene Symbol: ACACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009256.1 | NP_001009256.1 | 4 - 5 | 681 | 70 | |
| XM_027973950.1 | XP_027829751.1 | 3 - 4 | 977 | 70 | ||
| XM_027973951.1 | XP_027829752.1 | 3 - 4 | 667 | 70 | ||
| XM_027973952.1 | XP_027829753.1 | 2 - 3 | 554 | 70 | ||
| GenBank mRNA | AJ001056.1 | - | 2 - 3 | 559 | 70 | |
| X80045.1 | - | 2 | 681 | 70 | ||