Search
Search
| Gene Symbol: GNLY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105262.1 | NP_001098732.1 | 1 - 2 | 64 | 121 | |
| XM_027965710.1 | XP_027821511.1 | 1 - 2 | 121 | |||
| GenBank mRNA | EF654537.1 | - | 1 - 2 | 63 | 121 | |
| EF684914.1 | - | 1 - 2 | 64 | 121 | ||
Set Membership: |
|