Search
Search

XRRA1| Gene Symbol: XRRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096051.3 | XP_011951441.2 | 16 - 17 | 2167 | 59 | |
| XM_027979533.1 | XP_027835334.1 | 16 - 17 | 2072 | 59 | ||
| XM_027979534.1 | XP_027835335.1 | 15 - 16 | 2041 | 59 | ||
Set Membership: |
|