Search
Search

C21H11orf80| Gene Symbol: C21H11orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959697.1 | XP_027815498.1 | 12 - 13 | 1318 | 59 | |
| XM_027959698.1 | XP_027815499.1 | 11 - 12 | 1246 | 59 | ||
| XM_027959699.1 | XP_027815500.1 | 12 - 13 | 1240 | 59 | ||
| XM_027959700.1 | XP_027815501.1 | 10 - 11 | 1413 | 59 | ||
| XM_027959701.1 | XP_027815502.1 | 10 - 11 | 1084 | 59 | ||
| XM_027959702.1 | XP_027815503.1 | 10 - 11 | 1083 | 59 | ||
Set Membership: |
|