Search
Search

C21H11orf80| Gene Symbol: C21H11orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959697.1 | XP_027815498.1 | 7 - 8 | 595 | 68 | |
| XM_027959699.1 | XP_027815500.1 | 7 - 8 | 517 | 68 | ||
| XM_027959700.1 | XP_027815501.1 | 5 - 6 | 690 | 68 | ||
| XM_027959701.1 | XP_027815502.1 | 5 - 6 | 361 | 68 | ||
| XM_027959702.1 | XP_027815503.1 | 5 - 6 | 360 | 68 | ||
Set Membership: |
|