Search
Search

ASB11| Gene Symbol: ASB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021924.4 | XP_004021973.1 | 6 - 7 | 892 | 79 | |
| XM_004021925.4 | XP_004021974.1 | 6 - 7 | 896 | 79 | ||
| XM_012106253.2 | XP_011961643.1 | 8 - 9 | 942 | 79 | ||
| XM_012106254.2 | XP_011961644.1 | 8 - 9 | 937 | 79 | ||
| XM_012106256.3 | XP_011961646.1 | 6 - 7 | 857 | 79 | ||
| XM_027963638.1 | XP_027819439.1 | 5 - 6 | 787 | 79 | ||
| XM_027963639.1 | XP_027819440.1 | 5 - 6 | 758 | 79 | ||
| XM_027963640.1 | XP_027819441.1 | 5 - 6 | 821 | 79 | ||
Set Membership: |
|