Search
Search

IGF2BP3| Gene Symbol: IGF2BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007792.4 | XP_004007841.1 | 1 - 2 | 601 | 60 | |
| XM_012176514.3 | XP_012031904.1 | 1 - 2 | 600 | 60 | ||
Set Membership: |
|