Search
Search

ZNF385D| Gene Symbol: ZNF385D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962667.1 | XP_027818468.1 | 6 - 7 | 1233 | 62 | |
| XM_027962668.1 | XP_027818469.1 | 6 - 7 | 902 | 62 | ||
Set Membership: |
|